6-Methoxy-2-naphthaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

533084-5G
SIAL533084-5GEA 32.4 EUR
SIAL533084-5G
6-Methoxy-2-naphthaldehyd, Sigma-Aldrich®
6-Methoxy-2-naphthaldehyd
Formel: C₁₂H₁₀O₂
Molecular Weight: 186,21 g/mol
MDL: MFCD00081152
CAS-Nummer: 3453-33-6
EINECS: 222-377-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR