2-Chlorbenzaldehydoxim, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

533238-10G
SIAL533238-10GEA 112 EUR
SIAL533238-10G
2-Chlorbenzaldehydoxim, Sigma-Aldrich®
2-Chlorbenzaldehydoxim
Formel: C₇H₆ClNO
Molecular Weight: 155.58
MDL: MFCD00016406
CAS-Nummer: 3717-28-0
EINECS: 223-065-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR