2-Fluor-6-methylpyridin, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
2-Fluor-6-picolin
M-Clarity™ Qualitätsstufe = MQ100
533262-1G
SIAL533262-1GEA
26
EUR
SIAL533262-1G
2-Fluor-6-methylpyridin, Sigma-Aldrich®
2-Fluor-6-methylpyridin
Formel:
C₆H₆FN Molecular Weight: 111,12 g/mol |
MDL:
MFCD00041226 CAS-Nummer: 407-22-7 EINECS: 206-980-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...