4-Brom-2-methoxyphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromguajacol, 1-Brom-4-hydroxy-3-methoxybenzol, 5-Brom-2-hydroxyanisol

M-Clarity™ Qualitätsstufe = MQ200

533637-25G
SIAL533637-25GEA 300 EUR
SIAL533637-25G
4-Brom-2-methoxyphenol, Sigma-Aldrich®
4-Brom-2-methoxyphenol
Formel: C₇H₇BrO₂
Molecular Weight: 203,04 g/mol
MDL: MFCD00051937
CAS-Nummer: 7368-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR