5'-Brom-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Bromo-2-hydroxy-3-nitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

534153-25G
SIAL534153-25GEA 60.9 EUR
SIAL534153-25G
5'-Brom-2'-hydroxy-3'-nitroacetophenon, Sigma-Aldrich®
5'-Brom-2'-hydroxy-3'-nitroacetophenon
Formel: C₈H₆BrNO₄
Molecular Weight: 260,05 g/mol
MDL: MFCD01631134
CAS-Nummer: 70978-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR