4-(2-Methoxyethyl)phenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-(2-Methoxyethyl) phenol

M-Clarity™ Qualitätsstufe = MQ100

534595-25G
SIAL534595-25GEA 59.9 EUR
SIAL534595-25G
4-(2-Methoxyethyl)phenol, Sigma-Aldrich®
4-(2-Methoxyethyl)phenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00017537
CAS-Nummer: 56718-71-9
EINECS: 260-354-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR