2-Amino-5,6-dimethyl-4-hydroxypyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-5,6-dimethyl-4-pyrimidinol

M-Clarity™ Qualitätsstufe = MQ100

534617-1G-A
SIAL534617-1G-AEA 72 EUR
SIAL534617-1G-A
2-Amino-5,6-dimethyl-4-hydroxypyrimidin, Sigma-Aldrich®
2-Amino-5,6-dimethyl-4-hydroxypyrimidin
Formel: C₆H₉N₃O
Molecular Weight: 139,16 g/mol
MDL: MFCD00023258
CAS-Nummer: 3977-23-9
EINECS: 223-611-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR