(±)-4-Fluorstyroloxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

534765-5G
SIAL534765-5GEA 91.7 EUR
SIAL534765-5G
(±)-4-Fluorstyroloxid, Sigma-Aldrich®
(±)-4-Fluorstyroloxid
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
MDL: MFCD00043994
CAS-Nummer: 18511-62-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR