1-Phenyl-3-buten-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Phenyl-3-buten-1-ol

M-Clarity™ Qualitätsstufe = MQ100

535907-5G
SIAL535907-5GEA 113 EUR
SIAL535907-5G
1-Phenyl-3-buten-1-ol, Sigma-Aldrich®
4-Phenyl-1-buten-4-ol
Formel: C₁₀H₁₂O
Molecular Weight: 148,20 g/mol
MDL: MFCD00039617
CAS-Nummer: 936-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR