5-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

535982-5G
SIAL535982-5GEA 32.2 EUR
SIAL535982-5G
5-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®
5-Chlor-2-fluorbenzoesäure
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
MDL: MFCD00665762
CAS-Nummer: 394-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR