1-Iod-4-(trifluormethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluormethoxy)iodbenzol

M-Clarity™ Qualitätsstufe = MQ200

535990-5G
SIAL535990-5GEA 58.2 EUR
SIAL535990-5G
1-Iod-4-(trifluormethoxy)benzol, Sigma-Aldrich®
1-Iod-4-(trifluormethoxy)benzol
Formel: C₇H₄F₃IO
Molecular Weight: 288.01 g/mol
MDL: MFCD00042411
CAS-Nummer: 103962-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR