5-Chlor-2-fluoranilin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

536024-1ML
SIAL536024-1MLEA 50.3 EUR
SIAL536024-1ML
5-Chlor-2-fluoranilin, Sigma-Aldrich®
5-Chlor-2-fluoranilin
Formel: C₆H₅ClFN
Molecular Weight: 145,56 g/mol
MDL: MFCD00069416
CAS-Nummer: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR