5-Amino-2-mercaptobenzimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Amino-1H-benzoimidazole-2-thiol
M-Clarity™ Qualitätsstufe = MQ100
536067-25G
SIAL536067-25GEA
74.8
EUR
SIAL536067-25G
5-Amino-2-mercaptobenzimidazol, Sigma-Aldrich®
5-Amino-2-mercaptobenzimidazole
Formel:
C₇H₇N₃S |
MDL:
MFCD00022671 CAS-Nummer: 2818-66-8 EINECS: 220-574-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...