4'-Amino-3',5'-dichloracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4′-Amino-3′,5′-dichloroacetophenone

M-Clarity™ Qualitätsstufe = MQ100

536148-25G
SIAL536148-25GEA 102 EUR
SIAL536148-25G
4'-Amino-3',5'-dichloracetophenon, Sigma-Aldrich®
4'-Amino-3',5'-dichloracetophenon
Formel: C₈H₇Cl₂NO
Molecular Weight: 204,0500 g/mol
MDL: MFCD00238535
CAS-Nummer: 37148-48-4
EINECS: 253-368-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR