2-Fluorbenzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluorbenzohydrazid, 2-Fluorbenzol-1-carbohydrazid, 2-Fluorbenzoesäurehydrazid

M-Clarity™ Qualitätsstufe = MQ100

536288-10G
SIAL536288-10GEA 63.4 EUR
SIAL536288-10G
2-Fluorbenzhydrazid, Sigma-Aldrich®
2-Fluorbenzhydrazid
Formel: C₇H₇FN₂O
Molecular Weight: 154,14 g/mol
MDL: MFCD00025112
CAS-Nummer: 446-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR