3-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

536318-1G
SIAL536318-1GEA 31.2 EUR
SIAL536318-1G
3-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®
3-Chloro-2-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
MDL: MFCD00042506
CAS-Nummer: 161957-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR