3-Fluorbenzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluorbenzoesäurehydrazid, 3-Fluorbenzol-1-carbohydrazid, 3-Fluorbenzohydrazid

M-Clarity™ Qualitätsstufe = MQ100

536326-1G
SIAL536326-1GEA 22.5 EUR
SIAL536326-1G
3-Fluorbenzhydrazid, Sigma-Aldrich®
3-Fluorbenzhydrazid
Formel: C₇H₇FN₂O
Molecular Weight: 154,14 g/mol
MDL: MFCD00060561
CAS-Nummer: 499-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR