4-Fluorbenzhydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluorbenzol-1-carbohydrazid, 4-Fluorbenzohydrazid, 4-Fluorbenzoesäurehydrazid

M-Clarity™ Qualitätsstufe = MQ100

536334-5G
SIAL536334-5GEA 37.1 EUR
SIAL536334-5G
4-Fluorbenzhydrazid, Sigma-Aldrich®
4-Fluorbenzhydrazid
Formel: C₇H₇FN₂O
Molecular Weight: 154,14 g/mol
MDL: MFCD00060562
CAS-Nummer: 456-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR