2,7-Bis(brommethyl)naphthalin, Sigma-Aldrich®

Lieferant: Merck

53658-1G-F
SIAL53658-1G-FEA 468 EUR
SIAL53658-1G-F
2,7-Bis(brommethyl)naphthalin, Sigma-Aldrich®
2,7-Bis(brommethyl)naphthalin
Formel: C₁₂H₁₀Br₂
Molecular Weight: 314,158 g/mol
MDL: MFCD08277010
CAS-Nummer: 38309-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR