Biphenyl-4-sulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

536938-5G
SIAL536938-5GEA 47.6 EUR
SIAL536938-5G
Biphenyl-4-sulfonylchlorid, Sigma-Aldrich®
Biphenyl-4-sulfonylchlorid
Formel: C₁₂H₉ClO₂S
MDL: MFCD00078658
CAS-Nummer: 1623-93-4
EINECS: 216-609-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR