1-Brom-4-fluornaphthalin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

537195-5G
SIAL537195-5GEA 95.1 EUR
SIAL537195-5G
1-Brom-4-fluornaphthalin, Sigma-Aldrich®
1-Bromo-4-fluoronaphthalene
Formel: C₁₀H₆BrF
Molecular Weight: 225,06 g/mol
MDL: MFCD00051473
CAS-Nummer: 341-41-3
EINECS: 206-434-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR