6-Methoxy-2-naphthoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

537268-5G
SIAL537268-5GEA 57 EUR
SIAL537268-5G
6-Methoxy-2-naphthoesäure, Sigma-Aldrich®
6-Methoxy-2-naphthoesäure
Formel: C₁₂H₁₀O₃
MDL: MFCD00092750
CAS-Nummer: 2471-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR