Isobutylisobutyrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

537632-1L
SIAL537632-1LEA 140 EUR
SIAL537632-1L
Isobutylisobutyrat, Sigma-Aldrich®
Isobutylisobutyrat
Formel: C₈H₁₆O₂
Molecular Weight: 144,21 g/mol
MDL: MFCD00008916
CAS-Nummer: 97-85-8
EINECS: 202-612-5
UN: 2528
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR