(2-Fluor-3-(trifluormethyl)phenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Fluoro-3-(trifluoromethyl)phenylacetic acid

M-Clarity™ Qualitätsstufe = MQ100

538108-1G
SIAL538108-1GEA 55 EUR
SIAL538108-1G
(2-Fluor-3-(trifluormethyl)phenyl)essigsäure, Sigma-Aldrich®
(2-Fluor-3-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆F₄O₂
Molecular Weight: 222,14 g/mol
MDL: MFCD00061185
CAS-Nummer: 194943-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR