3'-(Trifluormethyl)propiophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-(Trifluormethyl)propiophenon

M-Clarity™ Qualitätsstufe = MQ100

538698-5G
SIAL538698-5GEA 58 EUR
SIAL538698-5G
3'-(Trifluormethyl)propiophenon, Sigma-Aldrich®
3'-(Trifluormethyl)propiophenon
Formel: C₁₀H₉F₃O
Molecular Weight: 202,18 g/mol
MDL: MFCD00039227
CAS-Nummer: 1533-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR