2'-(Trifluormethyl)propiophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: o-(Trifluormethyl)propiophenon

M-Clarity™ Qualitätsstufe = MQ100

538760-5G
SIAL538760-5GEA 37 EUR
SIAL538760-5G
2'-(Trifluormethyl)propiophenon, Sigma-Aldrich®
2'-(Trifluormethyl)propiophenon
Formel: C₁₀H₉F₃O
Molecular Weight: 202,18 g/mol
MDL: MFCD00013557
CAS-Nummer: 16185-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR