Ethylpentafluorpropionylacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl 3-oxo-4,4,5,5,5-pentafluoropentanoate
M-Clarity™ Qualitätsstufe = MQ100
538787-5G
SIAL538787-5GEA
89.7
EUR
SIAL538787-5G
Ethylpentafluorpropionylacetat, Sigma-Aldrich®
Ethylpentafluorpropionylacetat
Formel:
C₇H₇F₅O₃ Molecular Weight: 234,12 g/mol |
MDL:
MFCD00013569 CAS-Nummer: 663-35-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...