3-(Trifluormethyl)benzhydrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phenyl(3-(trifluoromethyl)phenyl)methanol

M-Clarity™ Qualitätsstufe = MQ100

538817-5G
SIAL538817-5GEA 61.2 EUR
SIAL538817-5G
3-(Trifluormethyl)benzhydrol, Sigma-Aldrich®
3-(Trifluormethyl)benzhydrol
Formel: C₁₄H₁₁F₃O
Molecular Weight: 252,23 g/mol
MDL: MFCD00014397
CAS-Nummer: 728-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR