Phenylsulfonylaceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Phenylsulfonyl)aceton

M-Clarity™ Qualitätsstufe = MQ200

539082-1G
SIAL539082-1GEA 0 EUR
SIAL539082-1G
Phenylsulfonylaceton, Sigma-Aldrich®
Phenylsulfonylaceton
Formel: C₉H₁₀O₃S
Molecular Weight: 198.24 g/mol
MDL: MFCD00047794
CAS-Nummer: 5000-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR