4-(Trifluormethyl)benzoesäurehydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide

M-Clarity™ Qualitätsstufe = MQ200

539171-1G
SIAL539171-1GEA 30.8 EUR
SIAL539171-1G
4-(Trifluormethyl)benzoesäurehydrazid, Sigma-Aldrich®
4-(Trifluoromethyl)benzoic acid hydrazide
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,15 g/mol
MDL: MFCD00051703
CAS-Nummer: 339-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR