Propyltriethoxysilan, Sigma-Aldrich®

Lieferant: Merck

539317-25ML
SIAL539317-25MLEA 0 EUR
SIAL539317-25ML
Propyltriethoxysilan, Sigma-Aldrich®
Propyltriethoxysilan
Formel: C₉H₂₂O₃Si
MDL: MFCD00026756
CAS-Nummer: 2550-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR