2-Fluor-6-(trifluormethyl)phenylacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

539635-1G
SIAL539635-1GEA 52 EUR
SIAL539635-1G
2-Fluor-6-(trifluormethyl)phenylacetonitril, Sigma-Aldrich®
2-Fluor-6-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
MDL: MFCD00061184
CAS-Nummer: 179946-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR