(R)-(+)-Epoxystyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

540099-5G
SIAL540099-5GEA 71 EUR
SIAL540099-5G
(R)-(+)-Epoxystyrol, Sigma-Aldrich®
(R)-(+)-Epoxystyrol
Formel: C₈H₈O
MDL: MFCD00066210
CAS-Nummer: 20780-53-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR