(2S)-(+)-2,3-Epoxypropyltoluol-4-sulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S)-(+)-Glycidyl tosylate, (s)-(+)-oxirane-2-methanol p-toluenesulphonate, (2S)-(+)-Glycidyl p-toluenesulphonate

M-Clarity™ Qualitätsstufe = MQ100

540129-5G
SIAL540129-5GEA 36.7 EUR
SIAL540129-5G
(2S)-(+)-2,3-Epoxypropyltoluol-4-sulfonat, Sigma-Aldrich®
(2S)-(+)-2,3-Epoxypropyltoluol-4-sulfonat
Formel: C₁₀H₁₂O₄S
Molecular Weight: 228.26 g/mol
MDL: MFCD00064489
CAS-Nummer: 70987-78-9
EINECS: 417-210-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR