Methyl-1-hydroxy-2-naphthoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Hydroxy-β-naphthalincarbonsäuremethylester, 1-Hydroxy-2-naphthoesäuremethylester, 1-Hydroxy-β-naphthoesäuremethylester, 1-Hydroxynaphthalin-2-carbonsäuremethylester, 1-Hydroxy-2-naphthalincarbonsäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

540188-5G
SIAL540188-5GEA 120 EUR
SIAL540188-5G
Methyl-1-hydroxy-2-naphthoat, Sigma-Aldrich®
Methyl-1-hydroxy-2-naphthoat
Formel: C₁₂H₁₀O₃
Molecular Weight: 202,21 g/mol
MDL: MFCD00043823
CAS-Nummer: 948-03-8
EINECS: 213-434-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR