Dimethyl-2,5-pyridindicarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl pyridine-2,5-dicarboxylate

M-Clarity™ Qualitätsstufe = MQ100

540234-25G
SIAL540234-25GEA 76.5 EUR
SIAL540234-25G
Dimethyl-2,5-pyridindicarboxylat, Sigma-Aldrich®
Dimethyl-2,5-pyridindicarboxylat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
MDL: MFCD00034767
CAS-Nummer: 881-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR