(R)-(-)-1,2-Propandiol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-1,2-Dihydroxypropan, (R)-(-)-1,2-Propylenglycol, (R)-(-)-1,2-Propylenglykol
MQ100
540242-25G
540242-5G
SIAL540242-25GEA
123
EUR
SIAL540242-25G
SIAL540242-5G
(R)-(-)-1,2-Propandiol, Sigma-Aldrich®
(R)-(-)-1,2-Propandiol
Formel:
C₃H₈O₂ Molecular Weight: 76.1 g/mol |
MDL:
MFCD00066248 CAS-Nummer: 4254-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...