(R)-(-)-1,2-Propandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-1,2-Dihydroxypropan, (R)-(-)-1,2-Propylenglycol, (R)-(-)-1,2-Propylenglykol

MQ100

540242-25G 540242-5G
SIAL540242-25GEA 123 EUR
SIAL540242-25G SIAL540242-5G
(R)-(-)-1,2-Propandiol, Sigma-Aldrich®
(R)-(-)-1,2-Propandiol
Formel: C₃H₈O₂
Molecular Weight: 76.1 g/mol
MDL: MFCD00066248
CAS-Nummer: 4254-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR