2,5-Dibrom-α,α,α-trifluortoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

M-Clarity™ Qualitätsstufe = MQ100

540277-5G
SIAL540277-5GEA 35.3 EUR
SIAL540277-5G
2,5-Dibrom-α,α,α-trifluortoluol, Sigma-Aldrich®
2,5-Dibrom-α,α,α-trifluortoluol
Formel: C₇H₃Br₂F₃
Molecular Weight: 303.9 g/mol
MDL: MFCD00013554
CAS-Nummer: 7657-09-2
EINECS: 231-620-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR