4-Iod-3-nitrotoluol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1-Iodo-4-methyl-2-nitrobenzene
M-Clarity™ Qualitätsstufe = MQ100
540447-5G
SIAL540447-5GEA
52
EUR
SIAL540447-5G
4-Iod-3-nitrotoluol, Sigma-Aldrich®
4-Iod-3-nitrotoluol
Formel:
C₇H₆INO₂ Molecular Weight: 263,03 g/mol Schmelzpunkt: 55 °C |
MDL:
MFCD00024196 CAS-Nummer: 5326-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...