3,4-Dimethylphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Dimethylphenylhydrazinhydrochlorid, 3,4-Dimethylphenylhydrazin Hydrochlorid, 1-(3,4-Dimethylphenyl)hydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

540455-25G
SIAL540455-25GEA 281 EUR
SIAL540455-25G
3,4-Dimethylphenylhydraziniumchlorid, Sigma-Aldrich®
3,4-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
MDL: MFCD00052270
CAS-Nummer: 60481-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR