Methyl-2,4-dichlorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2,4-dichlorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

540684-25G
SIAL540684-25GEA 94.7 EUR
SIAL540684-25G
Methyl-2,4-dichlorbenzoat, Sigma-Aldrich®
Methyl-2,4-dichlorbenzoat
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
MDL: MFCD00085117
CAS-Nummer: 35112-28-8
EINECS: 252-374-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR