Bis(cyclopentadienyl)zirkoniumdihydrid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

540692-1G
SIAL540692-1GEA 97.8 EUR
SIAL540692-1G
Bis(cyclopentadienyl)zirkoniumdihydrid, Sigma-Aldrich®
Bis(cyclopentadienyl)zirkoniumdihydrid
Formel: C₁₀H₁₂Zr
Molecular Weight: 223,43 g/mol
MDL: MFCD04126436
CAS-Nummer: 37342-98-6
UN: 3131
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR