(R)-(+)-3-Aminopyrrolidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-Pyrrolidin-3-amine
M-Clarity™ Qualitätsstufe = MQ100
540781-1G
SIAL540781-1GEA
226
EUR
SIAL540781-1G
(R)-(+)-3-Aminopyrrolidin, Sigma-Aldrich®
(R)-(+)-3-Aminopyrrolidin
Formel:
C₄H₁₀N₂ Molecular Weight: 86,14 g/mol |
MDL:
MFCD00143190 CAS-Nummer: 116183-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...