3-Brom-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromphenyl-methylcarbinol, 1-(3-Bromphenyl)ethanol, m-Brom-α-methylbenzylalkohol

M-Clarity™ Qualitätsstufe = MQ100

541028-25G
SIAL541028-25GEA 124 EUR
SIAL541028-25G
3-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
3-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00021863
CAS-Nummer: 52780-14-0
EINECS: 258-177-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR