Ethyl-2,6-dihydroxybenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

541079-5G
SIAL541079-5GEA 74 EUR
SIAL541079-5G
Ethyl-2,6-dihydroxybenzoat, Sigma-Aldrich®
Ethyl-2,6-dihydroxybenzoat
Formel: C₉H₁₀O₄
Molecular Weight: 182,18 g/mol
CAS-Nummer: 54640-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR