1-Chlor-3-fluor-4-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlor-1-fluor-2-iodbenzol, 4-Chlor-2-fluoriodbenzol

M-Clarity™ Qualitätsstufe = MQ100

541184-5G
SIAL541184-5GEA 40.3 EUR
SIAL541184-5G
1-Chlor-3-fluor-4-iodbenzol, Sigma-Aldrich®
1-Chloro-3-fluoro-4-iodobenzene
Formel: C₆H₃ClFI
Molecular Weight: 256,45 g/mol
MDL: MFCD00079729
CAS-Nummer: 6797-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR