1,2-Dichlor-4-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Dichloroiodobenzene

M-Clarity™ Qualitätsstufe = MQ100

541753-25G
SIAL541753-25GEA 87.5 EUR
SIAL541753-25G
1,2-Dichlor-4-iodbenzol, Sigma-Aldrich®
1,2-Dichlor-4-iodbenzol
Formel: C₆H₃Cl₂I
Molecular Weight: 272.9 g/mol
MDL: MFCD00019014
CAS-Nummer: 20555-91-3
EINECS: 243-874-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR