(±)-3,5-Difluormandelsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-3,5-Difluorphenylglykolsäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyethansäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyessigsäure

M-Clarity™ Qualitätsstufe = MQ100

541842-1G
SIAL541842-1GEA 82.6 EUR
SIAL541842-1G
(±)-3,5-Difluormandelsäure, Sigma-Aldrich®
(±)-3,5-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188,13 g/mol
MDL: MFCD00061300
CAS-Nummer: 132741-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR