Ammoniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

541893-5G
SIAL541893-5GEA 48.1 EUR
SIAL541893-5G
Ammoniumtetrafluoroborat, Sigma-Aldrich®
Ammonium tetrafluoroborate
Formel: H₄BF₄N
Molecular Weight: 104,84 g/mol
Schmelzpunkt: 510 °C
Dichte: 1,87 g/cm³ (20 °C)
MDL: MFCD06245785
CAS-Nummer: 13826-83-0
EINECS: 237-531-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR